Learn More
Mettler Toledo™ Polypropylene Titration Cups for Mettler Toledo Titrators
Product Code. 13102172 Shop All Mettler Toledo Products
Change view
Click to view available options
For Use With (Equipment):
Titrator
Unit Size:
Pack of 480
Product Code. For Use With (Equipment) Unit Size
13102172 Titrator Pack of 480
1 options
Product Code. 13102172 Supplier Mettler Toledo™ Supplier No. 51109389
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Accessory for use with Mettler Toledo Titrators.
For use with Mettler Toledo Titrators.
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Cup
For Use With (Application) Titration
Material Plastic
Volume (Metric) 100 mL
For Use With (Equipment) Titrator