missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Titration Beaker, for Mettler Toledo™ Titrators

Product Code. 10732152 Shop All Mettler Toledo Products
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Pack of 1400
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10732152 100 mL Pack of 1400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10732152 Supplier Mettler Toledo™ Supplier No. 00101974
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plastic beakers for use with titrators.

  • Material: Plastic
  • Capacity: 100mL
  • Quantity: 1400/Pk.
TRUSTED_SUSTAINABILITY

Specifications

Type Titration Beaker; 100mL; 1400/Cs.
For Use With (Application) Titration
Material Polypropylene
Volume (Metric) 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.