Learn More
Mettler Toledo™ Compact Stirrer, for Mettler Toledo™ Titrators
Product Code. 10700530 Shop All Mettler Toledo Products
Change view
Click to view available options
For Use With (Equipment):
DL15/22/28 titrators
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10700530 DL15/22/28 titrators Each
1 options
Product Code. 10700530 Supplier Mettler Toledo™ Supplier No. 51109150
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Accessory for use with DL22 Food and Beverage Titrator and DL15 and DL18 titrators.
Accessory for use with DL22 Food and Beverage Titrator and DL15 and DL18 titrators.
TRUSTED_SUSTAINABILITY
Specifications
Type Compact Overhead Stirrer with Two Propellers
For Use With (Application) Titration
Includes 2 Propeller stirrers
For Use With (Equipment) DL15/22/28 titrators