missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METTL7A Polyclonal Antibody

Description
A suggested positive control is A-20 cell lysate. PA5-20635 can be used with blocking peptide PEP-0755.
MettL7A belongs to the methyltransferase superfamily. It is a probable methyltransferase. Methyltransferase is a type of transferase enzyme which transfers a methyl group from a donor to an acceptor. Often methylation occurs on nucleic bases in DNA or amino acids in protein structures. DNA methylation is often utilized to silence and regulate genes without changing the original DNA sequence. DNA methylation may be necessary for normal growth from embryonic stages in mammals. When mutant embryonic stem cells lacking the murine DNA methyltransferase gene were introduced to a germline of mice they caused a recessive lethal phenotype. Methylation may also be linked to cancer development as methylation of tumor suppressor genes promotes tumorgenesis and metastasis.
Specifications
Specifications
| Antigen | METTL7A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | METTL7A |
| Gene Accession No. | Q9H8H3 |
| Gene Alias | 2210414H16Rik; 3300001H21Rik; Aam-B; methyltransferase like 7A; methyltransferase like 7A1; methyltransferase like 7A-like; methyltransferase-like protein 7A; METTL7A; Mettl7a1; PRO0066; Protein AAM-B; RGD1308407; UbiE1; UNQ1902/PRO4348 |
| Gene Symbols | METTL7A, Mettl7a1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?