missing translation for 'onlineSavingsMsg'
Learn More

METTL7A Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18407573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407573 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407573 Supplier Novus Biologicals Supplier No. NBP177336MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

METTL7A Polyclonal antibody specifically detects METTL7A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen METTL7A
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AAM-B, DKFZP586A0522, EC 2.1.1, EC 2.1.1.-, methyltransferase like 7A, methyltransferase-like protein 7A, Protein AAM-B
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide near the center of human MettL7A. The immunogen is located within amino acids 130 - 180 of MettL7A. Amino Acid Squence: SVKNQERILREV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25840
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.