Learn More
METTL5 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-16791-100UL
Beskrivning
The METTL5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human METTL5. This antibody recognizes human antigen. The METTL5 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifikationer
| METTL5 | |
| 1000 μg/mL | |
| FLJ10459, HSPC133, methyltransferase like 5, METTL5 | |
| 100 μL | |
| 29081 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.