Learn More
METTL5 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-16791-20UL
 View more versions of this product
Product Code. 18035555
  • £135.00 / 20µL
Description
The METTL5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human METTL5. This antibody recognizes human antigen. The METTL5 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
METTL5
1000 μg/mL
FLJ10459, HSPC133, methyltransferase like 5, METTL5
20 μL
29081
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human
150 kDa