missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METTL4 Recombinant Rabbit Monoclonal Antibody (9N4K4)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555486
This item is not returnable.
View return policy
Description
Immunogen Sequence: MSVVHQLSAG WLLDHLSFIN KINYQLHQHH EPCCRKKEFT TSVHFESLQM DSVSSSGVCA AFIASDSSTK PENDDGGNYE MFTRKFVFRP ELFDVTKPYI TPAVHKECQQ SNEKEDLMNG VKKEISISII GKKRKRCVVF NQGELDAMEY HTKIRELILD GSLQLIQEGL KSGFLYPLFE KQDKGSKPIT.
METTL4 gene ontology annotations related to this gene include methyltransferase activity; nucleic acid binding.
Specifications
| METTL4 | |
| Recombinant Monoclonal | |
| 1.18 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q3U034, Q8N3J2 | |
| METTL4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human METTL4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 9N4K4 | |
| Unconjugated | |
| METTL4 | |
| 2410198H06Rik; A730091E08Rik; AV296509; HsT661; methyltransferase like 4; methyltransferase-like protein 4; METTL4; N(6)-adenine-specific DNA methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; N(6)-adenine-specific methyltransferase METTL4; methyltransferase-like protein 4; RGD1306451 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 316731, 64863, 76781 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction