Learn More
METTL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15073-1-AP-150UL
 View more versions of this product
Product Code. 16874064
  • £349.00 / 150µL
Description
METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.
Specifications
Specifications
METTL3
Polyclonal
Unconjugated
METTL3
IME4, M6A, methyltransferase like 3, METTL3, MT A70, MTA70, Spo8
Rabbit
Antigen Affinity Chromatography
RUO
361035, 56335, 56339
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86U44, Q8C3P7
Mettl3
METTL3 Fusion Protein Ag7110
150 μL
Primary
Human, Mouse, Rat