missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METTL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145227
This item is not returnable.
View return policy
Description
Immunogen sequence: KEHCLVGVKG NPQGFNQGLD CDVIVAEVRS TSHKPDEIYG MIERLSPGTR KIELFGRPHN VQPNWITLGN QLDGIHLLDP DVVARFKQRY PDGII Antigen sequence identity: Mouse 100%; Rat 98%.
METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.
Specifications
| METTL3 | |
| Polyclonal | |
| Unconjugated | |
| METTL3 | |
| 2310024F18Rik; adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; fj82g07; hMETTL3; IME4; M6A; m6a methyltransferase; methyltransferase like 3; methyltransferase-like 3; methyltransferase-like protein 3; METTL3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MTA70; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit; N6-adenosine-methyltransferase subunit METTL3; Spo8; Unknown (protein for MGC:77093); wu:fj82g07; zgc:77093 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56339 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86U44 | |
| METTL3 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 480-574. Recombinant protein control fragment (Product #RP-109918). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction