missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 720347
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with Monkey, Horse, Bovine, Pig.
METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.Specifications
METTL3 | |
Polyclonal | |
Unconjugated | |
METTL3 | |
2310024F18Rik; adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; fj82g07; hMETTL3; IME4; M6A; m6a methyltransferase; methyltransferase like 3; methyltransferase-like 3; methyltransferase-like protein 3; METTL3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MTA70; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit; N6-adenosine-methyltransferase subunit METTL3; Spo8; Unknown (protein for MGC:77093); wu:fj82g07; zgc:77093 | |
Rabbit | |
Protein G | |
RUO | |
56335, 56339 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4 | |
Q86U44, Q8C3P7 | |
METTL3 | |
Protein corresponding to human METTL3 (aa280-aa580) | |
100μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |