Learn More
METTL25 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559478
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MCHYLKEERW CCGRNARMSA CLALERVAAG QGLPTESLFY RAVLQDIIKD CYGITKCDRH VGKIYSKCSS FLDYVRRSLK K Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
Putative methyltransferase.Specifications
METTL25 | |
Polyclonal | |
Unconjugated | |
METTL25 | |
C12orf26; methyltransferase like 25; methyltransferase-like protein 25; METTL25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84190 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6Q8 | |
METTL25 | |
Recombinant protein corresponding to Human METTL25 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only