missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL21C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546531
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AQQPGRRGEG LSSPGGWLEA EKKGAPQKDS TGGVLEESNK IEPSLHSLQK Sequence homology: Human: 100%
METTL21C Polyclonal antibody specifically detects METTL21C in Human samples. It is validated for Western Blot.Specifications
METTL21C | |
Polyclonal | |
Unconjugated | |
Mettl21c | |
A530098C11Rik; C13orf39; methyltransferase like 21C; methyltransferase-like protein 21C; METTL21C; Protein-lysine methyltransferase METTL21C; RGD1561425 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
196541 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VZV1 | |
Mettl21c | |
Synthetic peptide directed towards the N-terminal of human METTL21C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |