missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METTL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113304
This item is not returnable.
View return policy
Description
METTL2 Polyclonal Antibody for Western Blot, ELISA
METTL2A is probable methyltransferase.
Specifications
| METTL2 | |
| Polyclonal | |
| Unconjugated | |
| METTL2A | |
| HSPC266; methyltransferase like 2A; methyltransferase like 2B; methyltransferase-like protein 2A; Methyltransferase-like protein 2B; METL; METTL2; METTL2A; METTL2B; PSENIP1; tRNA N(3)-methylcytidine methyltransferase METTL2A; tRNA N(3)-methylcytidine methyltransferase METTL2B | |
| Rabbit | |
| Antigen affinity chromatography, Protein G | |
| RUO | |
| 339175 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.4 | |
| Q96IZ6 | |
| METTL2A | |
| Recombinant Human Methyltransferase-like protein 2A protein (1-242AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction