Learn More
METTL14 Antibody (CL4254) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59044JF646
Product Code. 30302326
  • £558.00 / 0.10mL
Description
METTL14 Monoclonal antibody specifically detects METTL14 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
METTL14
Monoclonal
Janelia Fluor 646
EC 2.1.1, EC 2.1.1.-, KIAA1627methyltransferase-like protein 14, methyltransferase like 14
This antibody was developed using a recombinant protein derived from Q9HCE5, with the exact immunogen sequence remaining proprietary.
0.1 mL
Lipid and Metabolism
57721
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL4254
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified