Learn More
METTL11B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA555753
Informations supplémentaires : Weight : 0.02000kg
Description
Immunogen sequence: GIIILKDNVA REGCILDLSD SSVTRDMDIL RSLIRKSGLV VLGQEKQDGF PEQCIPVWMF AL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Alpha-N-methyltransferase that methylates the N-terminus of target proteins containing the N-terminal motif [Ala/Pro/Ser]-Pro-Lys when the initiator Met is cleaved. Specifically catalyzes monomethylation of exposed alpha-amino group of Ala or Ser residue in the [Ala/Ser]-Pro-Lys motif and Pro in the Pro-Pro-Lys motif. May activate NTMT1 by priming its substrates for trimethylation.Spécification
METTL11B | |
Polyclonal | |
Unconjugated | |
Mettl11b | |
alpha N-terminal protein methyltransferase 1B; C1orf184; Gm207; HOMT1B; methyltransferase like 11B; methyltransferase-like protein 11B; METTL11B; NRMT2; NTM1B; RGD1564106; X-Pro-Lys N-terminal protein methyltransferase 1B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
149281 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VVY1 | |
Mettl11b | |
Recombinant protein corresponding to Human METTL11B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only