missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | METTL1 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16843954
|
Proteintech
14994-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16833954
|
Proteintech
14994-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is similar in sequence to the S. cerevisiae YDL201w gene. The gene product contains a conserved S-adenosylmethionine-binding motif and is inactivated by phosphorylation.Specifications
| METTL1 | |
| Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q9UBP6, Q9Z120 | |
| 17299, 4234, 679091 | |
| METTL1 Fusion Protein Ag6980 | |
| Primary | |
| -20°C | |
| METTL1 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C12orf1, methyltransferase like 1, METTL1, TRM8, tRNA(m7G46) methyltransferase, YDL201w | |
| Mettl1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title