missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL1 Polyclonal Antibody

METTL1 Polyclonal Antibody
Brand: Invitrogen PA565621
This item is not returnable.
View return policy
Description
METTL1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| METTL1 | |
| Polyclonal | |
| Unconjugated | |
| Mettl1 | |
| 2810012D02Rik; C12orf1; D1075-like gene product; hypothetical protein LOC449779; methyltransferase like 1; methyltransferase-like 1; methyltransferase-like protein 1; METTL1; miRNA (guanine-N(7)-)-methyltransferase; mRNA (guanine-N(7)-)-methyltransferase; TRM8; TRMT8; tRNA (guanine(46)-N(7))-methyltransferase; tRNA (guanine-N(7)-)-methyltransferase; tRNA (guanine-N(7)-)-methyltransferase-like protein; tRNA(m7G46)-methyltransferase; Unknown (protein for MGC:134433); YDL201w; zgc:103636 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4234 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBP6 | |
| Mettl1 | |
| Recombinant Human METTL1. Recombinant protein control fragment (Product #RP-104904). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction