missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22316
This item is not returnable.
View return policy
Description
METTL1 Polyclonal specifically detects METTL1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| METTL1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| C12orf1, D1075-like gene product, EC 2.1.1.33, FLJ95748, methyltransferase like 1, methyltransferase-like 1, Methyltransferase-like protein 1, TRM8, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase, YDL201w | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4234 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000 - 1:5000, Immunoprecipitation | |
| NP_005362.3 | |
| METTL1 | |
| The immunogen this antibody was made to, maps to a region between residue 175 to 225 of human Methyltransferase Like 1 using the numbering given in entry NP_005362.3 (GeneID 4234). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction