missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22316CL1
This item is not returnable.
View return policy
Description
METTL1 Polyclonal antibody specifically detects METTL1 in Human,Mouse samples. It is validated for Western Blot
Specifications
| METTL1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| C12orf1, D1075-like gene product, EC 2.1.1.33, FLJ95748, methyltransferase like 1, methyltransferase-like 1, Methyltransferase-like protein 1, TRM8, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase, YDL201w | |
| The immunogen this antibody was made to, maps to a region between residue 175 to 225 of human Methyltransferase Like 1 using the numbering given in entry NP_005362.3 (GeneID 4234). | |
| 0.1 mL | |
| Signal Transduction | |
| 4234 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction