missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole, MedChemExpress

Product Code. 16448448
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448448 10 g 10g
16430878 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448448 Supplier Medchem Express Supplier No. HYB0318
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Metronidazole is an orally active nitroimidazole antibiotic, can be used to research anaerobic infections. Metronidazole can cross blood brain barrier and results inflammation and skeletal muscle contraction under long-term application.

Metronidazole is an orally active nitroimidazole antibiotic, can be used to research anaerobic infections. Metronidazole can cross blood brain barrier and results inflammation and skeletal muscle contraction under long-term application.

Specifications

Chemical Name or Material Metronidazole
Physical Form Powder
CAS 443-48-1
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C6H9N3O3
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 35 mg/mL (204.50 mM; Need ultrasonic and warming) ∣H2O : 16.67 mg/mL (97.40 mM; Need ultrasonic)
SMILES OCCN1C([N+]([O-])=O)=CN=C1C
Molecular Weight (g/mol) 171.15
Formula Weight 171.15
Percent Purity 98.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.