missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole D4 (ethylene D4) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30320389
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30320389 1 mL 1mL
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30320389

Brand: Dr. Ehrenstorfer DREA15201001AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Metronidazole D4 (ethylene D4)
CAS 1261392-47-5
Solvent or Matrix Acetonitrile
Molecular Formula C6 2H4 H5 N3 O3
Quantity 1 mL
InChI Formula InChI=1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3/i2D2,3D2
SMILES [2H]C([2H])(O)C([2H])([2H])n1c(C)ncc1[N+](=O)[O-]
IUPAC Name 1,1,2,2-tetradeuterio-2-(2-methyl-5-nitroimidazol-1-yl)ethanol
Molecular Weight (g/mol) 175.18
Formula Weight 175.0895 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.