missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole Beta-D-Glucuronide, TRC

Product Code. 30478508
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478508 1 mg 1mg
30476517 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478508 Supplier Toronto Research Chemicals Supplier No. TRCM3388901MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100495-98-5
Molecular Formula C12 H17 N3 O9
Molecular Weight (g/mol) 347.28
Synonym Metronidazole-β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-[2-(2-methyl-5-nitroimidazol-1-yl)ethoxy]oxane-2-carboxylic acid
SMILES Cc1ncc([N+](=O)[O-])n1CCO[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C12H17N3O9/c1-5-13-4-6(15(21)22)14(5)2-3-23-12-9(18)7(16)8(17)10(24-12)11(19)20/h4,7-10,12,16-18H,2-3H2,1H3,(H,19,20)/t7-,8-,9+,10-,12+/m0/s1
Formula Weight 347.0965
Chemical Name or Material Metronidazole beta-D-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.