missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MetroMax i™ Lab Worktable Accessory, Drawer

The Metro MetroMax i™ Lab Worktable Drawer comes in a choice of sizes to provide extra room and functionality in your storage setup.

Brand:  Metro™ LTD6L

Additional Details : Weight : 4.35200kg

 View more versions of this product

Product Code. 15937061

  • £550.00 / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket

Description

Description

  • Drawers fit 30, 36, 48 and 60 in. (76, 91.4, 121.9, and 152.4cm) table lengths
  • Locking feature available

Specifications

Specifications

Locking Drawer
6 in.
15.2 cm
For 30, 36, 48 and 60 in. lab worktables
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers