missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MetroMax i Glassware Cart, Accessories

Product Code. 11735659
Change view
Click to view available options
Product Type:
Glassware Transport Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11735659 Glassware Transport Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11735659 Supplier Metro™ Supplier No. GWOLYKIT
  • ONLINE EXCLUSIVE PRICE
    £299.00
    £205.32 / Pack of 1
    Save £93.68
    31% Off
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with MetroMax i carts

  • 36 in. basket for glassware cart

Specifications

Product Type Glassware Transport Kit
Color Gray
For Use With (Application) For easy and efficient way to transport laboratory Glassware
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.