missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Lab Worktable Accessory, Backsplash

Match length and color with selected countertop

Brand:  Metro™ LT30BSB

Additional Details : Weight : 1.99200kg

 View more versions of this product

Product Code. 15907021

  • £104.00 / Each
Estimated Shipment: 09-07-2024
to see stock.
Add to basket

Description

Description

  • 4 in. high (10.2 cm)
  • Attaches to phenolic table top using pre-drilled holes

Specifications

Specifications

27.75 in.
4 in.
Black
10.2 cm
70.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers