missing translation for 'onlineSavingsMsg'
Learn More

Metoxuron, TRC

Product Code. 30481028
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481028 25 mg 25mg
30476953 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481028 Supplier Toronto Research Chemicals Supplier No. TRCM33884325MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19937-59-8
Molecular Formula C10 H13 Cl N2 O2
Molecular Weight (g/mol) 228.68
Synonym Urea, 3-(3-chloro-4-methoxyphenyl)-1,1-dimethyl- (6CI,8CI), N'-(3-Chloro-4-methoxyphenyl)-N,N-dimethylurea, 1-(3-Chloro-4-methoxyphenyl)-3,3-dimethylurea, 3-(3-Chloro-4-methoxyphenyl)-1,1-dimethylurea, Dosanex, Metoxuran, Metoxuron, N-(3-Chloro-4-methoxyphenyl)-N',N'-dimethylurea, Purivel, SAN 6602, SAN 7102, Urea, N'-(3-chloro-4-methoxyphenyl)-N,N-dimethyl-
IUPAC Name 3-(3-chloro-4-methoxyphenyl)-1,1-dimethylurea
SMILES COc1ccc(NC(=O)N(C)C)cc1Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H13ClN2O2/c1-13(2)10(14)12-7-4-5-9(15-3)8(11)6-7/h4-6H,1-3H3,(H,12,14)
Formula Weight 228.0666
Chemical Name or Material Metoxuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.