missing translation for 'onlineSavingsMsg'
Learn More

Metoprolol tartrate, Dr. Ehrenstorfer

Product Code. 30324493
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324493 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324493 Supplier Dr. Ehrenstorfer Supplier No. DREC15176020100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Metoprolol Tartrate
CAS 56392-17-7
Analyte or Component Names Metoprolol Tartrate
Shipping Condition +20°C
Molecular Formula 2 C15 H25 N O3 . C4 H6 O6
Certifications/Compliance ISO 17034
Synonym Metoprolol Tartrate, 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-, (2R,3R)-2,3-dihydroxybutanedioate (2:1), 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-, (2R,3R)-2,3-dihydroxybutanedioate (2:1) (salt) (9CI), 1-(Isopropylamino)-3-[4-(2-methoxyethyl)phenoxy]-2-propanol tartrate, 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (2:1) (salt), Beloc, Betaloc, CGP 2175E, H 93/26, HCTCGP 2175E, Lopresor, Metapro, Metoprolol (+)-tartrate, Metoprolol tartrate (2:1), Prelis, Seloken, Selopral, Slow Lopresor, Vasocardin, Lapressor, 1-[4-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol Hemi (+)-Tartrate, Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-, (+/-)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (2:1) (salt), (±)-1-(Isopropylamino)-3-[4-(2-methoxyethyl)phenoxy]-2-propanol tartrate, (±)-Metoprolol bitartrate, (±)-Metoprolol tartrate, Metoprolol tartrate
InChI Formula InChI=1S/2C15H25NO3.C4H6O6/c2*1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;5-1(3(7)8)2(6)4(9)10/h2*4-7,12,14,16-17H,8-11H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1
SMILES COCCc1ccc(OCC(O)CNC(C)C)cc1.COCCc2ccc(OCC(O)CNC(C)C)cc2.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molecular Weight (g/mol) 684.81
Quantity 100 mg
Formula Weight 684.3833 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.