missing translation for 'onlineSavingsMsg'
Learn More

Metolazone, TRC

Product Code. 30133468
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133468 25 mg 25mg
30010224 5 mg 5mg
30006384 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133468 Supplier Toronto Research Chemicals Supplier No. TRCM33875025MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17560-51-9
Molecular Formula C16 H16 Cl N3 O3 S
Molecular Weight (g/mol) 365.83
Synonym Metolazone, (2RS)-7-Chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2,3,4-tetrahydroquinazoline-6-sulfonamide, 6-Quinazolinesulfonamide, 7-chloro-1,2,3,4-tetrahydro-2-methyl-3-(2-methylphenyl)-4-oxo- (9CI, ACI), 6-Quinazolinesulfonamide, 7-chloro-1,2,3,4-tetrahydro-2-methyl-4-oxo-3-o-tolyl- (8CI), 7-Chloro-1,2,3,4-tetrahydro-2-methyl-3-(2-methylphenyl)-4-oxo-6-quinazolinesulfonamide (ACI), 2-Methyl-3-(o-tolyl)-6-sulfamyl-7-chloro-1,2,3,4-tetrahydro-4-quinazolinone, 7-Chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2-dihydroquinazoline-6-sulfonamide, Diulo, MeSH ID: D008788, Metazoline, Metenix, Metozalone, Mykrox, Normelan, Oldren, SR 720-22, Xuret, Zaroxolyn
IUPAC Name 7-chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2-dihydroquinazoline-6-sulfonamide
SMILES CC1Nc2cc(Cl)c(cc2C(=O)N1c3ccccc3C)S(=O)(=O)N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H16ClN3O3S/c1-9-5-3-4-6-14(9)20-10(2)19-13-8-12(17)15(24(18,22)23)7-11(13)16(20)21/h3-8,10,19H,1-2H3,(H2,18,22,23)
Formula Weight 365.0601
Chemical Name or Material Metolazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.