missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metolazone, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Metolazone (SR-720-22) is primarily used to treat congestive heart failure and high blood pressure.
Specifications
Specifications
| Chemical Name or Material | Metolazone |
| CAS | 17560-51-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H16ClN3O3S |
| Quantity | 100 mg |
| Synonym | SR-720-22 |
| Solubility Information | DMSO : ≥ 100 mg/mL (273.35 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=S(C1=CC2=C(NC(C)N(C3=CC=CC=C3C)C2=O)C=C1Cl)(N)=O |
| Molecular Weight (g/mol) | 365.83 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction