missing translation for 'onlineSavingsMsg'
Learn More

Metoclopramide Hydrochloride, TRC

Product Code. 30137324
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137324 10 g 10g
30132867 1 g 1g
30003224 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137324 Supplier Toronto Research Chemicals Supplier No. TRCM33868510G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7232-21-5
Molecular Formula C14 H22 Cl N3 O2 . Cl H
Molecular Weight (g/mol) 336.26
Synonym Benzamide, 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxy-, hydrochloride (1:1) (ACI), Benzamide, 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxy-, monohydrochloride (9CI), o-Anisamide, 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-, monohydrochloride (8CI), 4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-o-anisamide hydrochloride, AHR 3070-C, Ami-metoclopramide, Cerucal, Maxolon, Metoclopramide hydrochloride, Metoclopramide monohydrochloride, Metoclopramide-Hakim, Metpamid, Mexeron, NSC 354467, Paspertin, Perinorm, Plazilin, Primperan, Primperan (tablet), Reglan, Rimetin, Telperane, Tserulan
IUPAC Name 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide;hydrochloride
SMILES Cl.CCN(CC)CCNC(=O)c1cc(Cl)c(N)cc1OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H22ClN3O2.ClH/c1-4-18(5-2)7-6-17-14(19)10-8-11(15)12(16)9-13(10)20-3;/h8-9H,4-7,16H2,1-3H3,(H,17,19);1H
Formula Weight 335.1167
Chemical Name or Material Metoclopramide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.