missing translation for 'onlineSavingsMsg'
Learn More

Metoclopramide hydrochloride

Product Code. 15494129
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494129 25 g 25g
15484129 10 g 10g
15404139 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15494129 Supplier Thermo Scientific Alfa Aesar Supplier No. J61545.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-HT3 serotonin antagonist; D2 dopamine antagonist | CAS: 54143-57-6 | C14H25Cl2N3O3 | 354.27 g/mol

Metoclopramide hydrochloride is used as a digestive system drugs. It is used for anti-emetic, used for digestive dysfunction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metoclopramide hydrochloride is used as a digestive system drugs. It is used for anti-emetic, used for digestive dysfunction.

Solubility
It is very soluble in water, easily soluble in ethyl alcohol, slightly soluble in dichloromethane.

Notes
Hygroscopic, Refrigerator, Under Inert atmosphere. Incompatible to strong alkalis, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54143-57-6
Molecular Formula C14H25Cl2N3O3
Molecular Weight (g/mol) 354.27
MDL Number MFCD00150634
InChI Key KJBLQGHJOCAOJP-UHFFFAOYSA-N
PubChem CID 441347
ChEBI CHEBI:6899
IUPAC Name 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride
SMILES O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC

Specifications

Color White to Yellow
Quantity 25 g
Sensitivity Light sensitive
Merck Index 14,6143
Solubility Information It is very soluble in water,easily soluble in ethyl alcohol,slightly soluble in dichloromethane.
Formula Weight 336.26
Physical Form Crystalline Solid
Chemical Name or Material Metoclopramide hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.