Learn More
METL5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558301
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DIDEDALEIF NRNAEEFELT NIDMVQCDVC LLSNRMSKSF DTVIMNPPFG T Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
Catalytic subunit of a heterodimer with TRMT112, which specifically methylates the 6th position of adenine in position 1832 of 18S rRNA. [UniProt]Specifications
METL5 | |
Polyclonal | |
Unconjugated | |
METTL5 | |
2810410A08Rik; DC3; HSPC133; im:7137598; methyltransferase like 5; methyltransferase-like protein 5; Mettl5; RGD1566062; rRNA N6-adenosine-methyltransferase METTL5; rRNA N6-adenosine-methyltransferase METTL5; methyltransferase-like protein 5; zgc:103655 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29081 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRN9 | |
METTL5 | |
Recombinant protein corresponding to Human METL5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only