missing translation for 'onlineSavingsMsg'
Learn More

Metiamide, MedChemExpress

Product Code. 16489177
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489177

Brand: Medchem Express HY15540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Metiamide (SK&F 92058) is a histamine H2-receptor antagonist developed from another H2 antagonist, burimamide.

Metiamide (SK&F 92058) is a histamine H2-receptor antagonist developed from another H2 antagonist, burimamide.

Specifications

Chemical Name or Material Metiamide
Physical Form Solid
CAS 34839-70-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C9H16N4S2
Synonym SK&F 92058
Solubility Information DMSO : ≥ 2.4 mg/mL (9.82 mM)
SMILES S=C(NCCSCC1=C(C)N=CN1)NC
Molecular Weight (g/mol) 244.38
Formula Weight 244.38
Percent Purity 96.39%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.