missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Methysergide maleate
GREENER_CHOICE

Product Code. 30001187
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
30001187 10 mg 10mg
18742764 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 30001187

Brand: Tocris Bioscience™ 1064/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-HT1/5-HT2 antagonist

Mixed 5-HT1/5-HT2 receptor antagonist.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-49-7
Molecular Formula C25H31N3O6

Specifications

Quantity 10 mg
Formula Weight 469.54
Purity >99%
Molecular Weight (g/mol) 469.54
Chemical Name or Material Methysergide maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.