accessibility menu, dialog, popup

UserName

CAS RN 129-49-7

CH3(S)OHNHO(R)NCH3(R)NH3COHOOHO(Z)

CAS RN 129-49-7

IUPAC Name: (2Z)-but-2-enedioic acid; (4R,7R)-N-[(2S)-1-hydroxybutan-2-yl]-6,11-dimethyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),2,9,12,14-pentaene-4-carboxamide
Synonyms: maleic acid; methysergide
Molecular Weight (g/mol): 469.54
Molecular Formula: C25H31N3O6
InChi Key: LWYXFDXUMVEZKS-ZVFOLQIPSA-N
SMILES: OC(=O)\C=C/C(O)=O.CC[C@@H](CO)NC(=O)[C@H]1CN(C)[C@@H]2CC3=CN(C)C4=CC=CC(=C34)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results