missing translation for 'onlineSavingsMsg'
Learn More

Methylzinc chloride, 2M solution in THF, AcroSeal™

Product Code. 11955981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11955981 50 mL AcroSeal™ Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11955981 Supplier Thermo Scientific Acros Supplier No. 436080500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5158-46-3 | CH3ClZn | 115.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5158-46-3, 109-99-9
Color Colorless to Yellow
CAS Min % 21.31, 75.32
CAS Max % 24.68, 78.69
Physical Form Turbid Solution
Quantity 50 mL
Molecular Formula CH3ClZn
MDL Number MFCD00192334
Flash Point −17°C
Synonym methylzinc chloride, chloro methyl zinc, methylzinc chloride solution, methylzinc chloride solution, 2.0 m in thf
InChI Key NMLXKNNXODLJIN-UHFFFAOYSA-M
SMILES [CH3-].Cl[Zn+]
IUPAC Name carbanide;chlorozinc(1+)
Molecular Weight (g/mol) 115.87
PubChem CID 4152418
Formula Weight 115.87
Concentration or Composition (by Analyte or Components) 1.90 to 2.20M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.033
Density 1.0330g/mL
Chemical Name or Material Methylzinc chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylzinc chloride, 2M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.