missing translation for 'onlineSavingsMsg'
Learn More

Methylurea, 98%

Product Code. 11345045
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345045 250 g 250g
11335045 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345045 Supplier Thermo Scientific Alfa Aesar Supplier No. B24772.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-50-5 | C2H6N2O | 74.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-50-5
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.083
MDL Number MFCD00007950
InChI Key XGEGHDBEHXKFPX-UHFFFAOYSA-N
Synonym 1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
PubChem CID 11719
ChEBI CHEBI:44383
IUPAC Name methylurea
SMILES CNC(=O)N

Specifications

Melting Point 96°C to 101°C
pH 6.7
Odor Odorless
Quantity 250 g
Beilstein 878189
Formula Weight 74.08
Percent Purity 98%
Chemical Name or Material Methylurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.