missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylsilane, 97%

Product Code. 11363126
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363126 25 g 25g
11343126 1 g 1g
11353126 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363126 Supplier Thermo Scientific Alfa Aesar Supplier No. L10995.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 791-29-7 | C19H18Si | 274.438 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 791-29-7
Molecular Formula C19H18Si
Molecular Weight (g/mol) 274.438
MDL Number MFCD00041325
InChI Key GIGVICQLYWGMGW-UHFFFAOYSA-N
Synonym silane, methyltriphenyl, triphenylmethylsilane, methyl triphenyl silane, methyl-tri phenyl silane, methyltriphenylsilane
PubChem CID 522714
IUPAC Name methyl(triphenyl)silane
SMILES C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 66°C to 68°C
Quantity 25 g
Beilstein 2940360
Formula Weight 274.44
Percent Purity 97%
Chemical Name or Material Methyltriphenylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.