missing translation for 'onlineSavingsMsg'
Learn More

Methyltriethoxysilane, 98+%

Product Code. 10236230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10236230 250 mL Glass bottle 250mL
10276180 25 mL Glass bottle 25mL
13410887 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10236230 Supplier Thermo Scientific Acros Supplier No. 174622500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2031-67-6 | C7H18O3Si | 178.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2031-67-6
Molecular Formula C7H18O3Si
Molecular Weight (g/mol) 178.30
MDL Number MFCD00009066
InChI Key CPUDPFPXCZDNGI-UHFFFAOYSA-N
Synonym methyltriethoxysilane, triethoxy methyl silane, methyl triethoxysilane, silane, triethoxymethyl, triethoxysilylmethane, union carbide a-162, ici-ep 5850, silane, methyltriethoxy, methaneorthosiliconic acid, triethyl ester, mtes
PubChem CID 16241
IUPAC Name triethoxy(methyl)silane
SMILES CCO[Si](C)(OCC)OCC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -40.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 141.0°C to 143.0°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3Si(OC2H5)3
Refractive Index 1.3825 to 1.3845
Quantity 250 mL
Beilstein 04,629
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Viscosity 0.6 mPa.s (20°C)
Formula Weight 178.3
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyltriethoxysilane, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.