missing translation for 'onlineSavingsMsg'
Learn More

Methyltriethoxysilane, 98%

Product Code. 11336776
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336776 1000 mL 1 L
11326776 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336776 Supplier Thermo Scientific Alfa Aesar Supplier No. L04059.AU
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2031-67-6 | C7H18O3Si | 178.30 g/mol

Methyltriethoxysilane is used in production of silicone polymers or silicone resin. It is used in non-metal surface treatment, non-aqueous polymer preparation, manufacture of semi-conductors, coatings and sealants. It finds application as cross linking agent and an adhesion promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltriethoxysilane is used in production of silicone polymers or silicone resin. It is used in non-metal surface treatment, non-aqueous polymer preparation, manufacture of semi-conductors, coatings and sealants. It finds application as cross linking agent and an adhesion promoter.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2031-67-6
Molecular Formula C7H18O3Si
Molecular Weight (g/mol) 178.30
MDL Number MFCD00009066
InChI Key CPUDPFPXCZDNGI-UHFFFAOYSA-N
Synonym methyltriethoxysilane, triethoxy methyl silane, methyl triethoxysilane, silane, triethoxymethyl, triethoxysilylmethane, union carbide a-162, ici-ep 5850, silane, methyltriethoxy, methaneorthosiliconic acid, triethyl ester, mtes
PubChem CID 16241
IUPAC Name triethoxy(methyl)silane
SMILES CCO[Si](C)(OCC)OCC

Specifications

Boiling Point 141°C to 143°C
Melting Point <-50°C
Quantity 1000 mL
Linear Formula CH3Si(OCH2CH3)3
UN Number UN1993
Beilstein 1742453
Flash Point 23°C (73°F)
Solubility Information Immiscible with water.
Formula Weight 178.3
Percent Purity 98%
Odor Mild
Refractive Index 1.3835
Sensitivity Moisture sensitive
Density 0.897
Chemical Name or Material Methyltriethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyltriethoxysilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methyltriethoxy-silane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.