missing translation for 'onlineSavingsMsg'
Learn More

Methyltrichlorosilane, 97%

Product Code. 11486410
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11486410

Brand: Thermo Scientific Alfa Aesar B23107.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-79-6 | CH3Cl3Si | 149.47 g/mol

Methyltrichlorosilane is used in production of methyl silicone resins, its vapor reacts with water on surfaces to give a thin layer of methylpolysiloxane which make it a water-repellent film. A combination of methyltrichlorosilane and sodium iodide can be used to cleave carbon-oxygen bonds such as methyl ethers. It is used as a precursor for forming various cross-linked siloxane polymers. Trichloromethylsilane is the starting material for the production of pure silicon for manufacture of semiconductors and optical fibers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltrichlorosilane is used in production of methyl silicone resins, its vapor reacts with water on surfaces to give a thin layer of methylpolysiloxane which make it a water-repellent film. A combination of methyltrichlorosilane and sodium iodide can be used to cleave carbon-oxygen bonds such as methyl ethers. It is used as a precursor for forming various cross-linked siloxane polymers. Trichloromethylsilane is the starting material for the production of pure silicon for manufacture of semiconductors and optical fibers.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-79-6
Molecular Formula CH3Cl3Si
Molecular Weight (g/mol) 149.47
MDL Number MFCD00000481
InChI Key JLUFWMXJHAVVNN-UHFFFAOYSA-N
Synonym methyltrichlorosilane, trichloro methyl silane, silane, trichloromethyl, methyl-trichlorsilan, methylsilicochloroform, trichloromethylsilicon, methylsilyl trichloride, monomethyltrichlorosilane, silane, methyltrichloro, trichlor-methylsilan
PubChem CID 6399
IUPAC Name trichloro(methyl)silane
SMILES C[Si](Cl)(Cl)Cl

Specifications

Boiling Point 64°C to 66°C
Melting Point -78°C
Quantity 100 g
UN Number UN1250
Beilstein 1361381
Flash Point 8°C (46°F)
Solubility Information Reacts with water.
Formula Weight 149.48
Percent Purity 97%
Odor Pungent
Refractive Index 1.4115
Sensitivity Moisture sensitive
Density 1.27
Chemical Name or Material Methyltrichlorosilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyltrichlorosilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methyltrichlorosilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.