missing translation for 'onlineSavingsMsg'
Learn More

Methyltri-n-butylammonium chloride, Aliquat 175 (75% aq. soln.)

Product Code. 11314885
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
This item is not returnable. View return policy

Product Code. 11314885

Brand: Thermo Scientific Alfa Aesar L18019.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56375-79-2 | C13H30ClN | 235.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56375-79-2
Molecular Formula C13H30ClN
Molecular Weight (g/mol) 235.84
MDL Number MFCD00011847
InChI Key IPILPUZVTYHGIL-UHFFFAOYSA-M
Synonym tributylmethylammonium chloride, n,n-dibutyl-n-methylbutan-1-aminium chloride, methyltributylammonium chloride, methyl tributyl ammonium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride, unii-az0b34me3u, az0b34me3u, tributyl methyl azanium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride 1:1, acmc-20aker
PubChem CID 91822
IUPAC Name tributyl(methyl)azanium;chloride
SMILES CCCC[N+](C)(CCCC)CCCC.[Cl-]

Specifications

Melting Point -33°C
Density 0.964
Boiling Point 84°C to 85°C (101 mmHg)
Quantity 100 g
Beilstein 6300212
Sensitivity Hygroscopic
Formula Weight 235.84
Concentration or Composition (by Analyte or Components) 75% aq. soln.
Chemical Name or Material Methyltri-n-butylammonium chloride, Aliquat 175

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.