missing translation for 'onlineSavingsMsg'
Learn More

Methyltin Trichloride, TRC

Product Code. 30480129
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30480129 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480129 Supplier Toronto Research Chemicals Supplier No. TRCM331130500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trichloromethylstannane
CAS 993-16-8
Molecular Formula C H3 Cl3 Sn
Quantity 500 mg
Synonym Stannane, trichloromethyl-, Methyltin trichloride (6CI), Trichloromethylstannane, Methyltrichlorostannane, Methyltrichlorotin, Monomethyltin trichloride, Trichloromethyltin
InChI Formula InChI=1S/CH3.3ClH.Sn/h1H3;3*1H;/q;;;;+3/p-3
SMILES C[Sn](Cl)(Cl)Cl
IUPAC Name trichloro(methyl)stannane
Molecular Weight (g/mol) 240.1
Formula Weight 239.8322
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.