missing translation for 'onlineSavingsMsg'
Learn More

Methylthymol Blue Sodium Salt (Technical Grade), TRC

Product Code. 30471556
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471556 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471556 Supplier Toronto Research Chemicals Supplier No. TRCM3386551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylthymol Blue Sodium Salt
CAS 1945-77-3
Molecular Formula C37H40N2Na4O13S
Quantity 1 g
Synonym 3H-2,1-Benzoxathiole Glycine Derivative, S,S-dioxide-N,N'-[3H-2,1-benzoxathiol-3-ylidenebis[[6-hydroxy-2-methyl-5-(1-methylethyl)-3,1-phenylene]methylene]]bis[N-(carboxymethyl)glycine Tetrasodium Salt, N,N'-[(1,1-Dioxido-3H-2,1-benzoxathiol-3-ylidene)bis[[6-hydroxy-2-methyl-5-(1-methylethyl)-3,1-phenylene]methylene]]bis[N-(carboxymethyl)glycine Tetrasodium Salt
InChI Formula InChI=1S/C37H44N2O13S.4Na/c1-19(2)23-11-28(21(5)25(35(23)48)13-38(15-31(40)41)16-32(42)43)37(27-9-7-8-10-30(27)53(50,51)52-37)29-12-24(20(3)4)36(49)26(22(29)6)14-39(17-33(44)45)18-34(46)47;;;;/h7-12,19-20,48-49H,13-18H2,1-6H3,(H,40,41)(H,42,43)(H,44,45)(H,46,47);;;;/q;4*+1/p-4
SMILES O=C(CN(CC1=C(C)C(C2(C3=CC(C(C)C)=C(C(CN(CC(O[Na])=O)CC(O[Na])=O)=C3C)O)OS(=O)(C4=C2C=CC=C4)=O)=CC(C(C)C)=C1O)CC(O[Na])=O)O[Na]
IUPAC Name sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetate
Molecular Weight (g/mol) 844.74
Formula Weight 844.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.