missing translation for 'onlineSavingsMsg'
Learn More

Methylthymol Blue Sodium Salt, Honeywell Fluka™

Product Code. 15643170 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15643170 1 g Glass Bottle 1g
15653170 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15643170 Supplier Honeywell-Fluka Supplier No. 310421G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1945-77-3
Molecular Formula C37H44N2Na4O13S
Molecular Weight (g/mol) 848.779
MDL Number MFCD00148905
InChI Key LGVVZVZPOQJZKT-UHFFFAOYSA-N
PubChem CID 131850582
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]

Specifications

Linear Formula C37H40N2O13SNa4
UN Number NONH for all modes of transport
Formula Weight 844.74g/mol
Packaging Glass Bottle
Quantity 1 g
Chemical Name or Material Methylthymol Blue Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.