missing translation for 'onlineSavingsMsg'
Learn More

Methylthiouracil, MedChemExpress

Product Code. 16474377
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474377 50 mg 50mg
16468346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16474377 Supplier Medchem Express Supplier No. HYB0513
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Methylthiouracil is an antithyroid agent. Methylthiouracil suppresses the production TNF-α and IL-6, and the activation of NF-κB and ERK1/2.

Methylthiouracil is an antithyroid agent. Methylthiouracil suppresses the production TNF-α and IL-6, and the activation of NF-κB and ERK1/2.

Specifications

Chemical Name or Material Methylthiouracil
Physical Form Solid
CAS 56-04-2
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C5H6N2OS
For Use With (Application) COVID-19-immunoregulation
Synonym MTU
Solubility Information DMSO : ≥ 50 mg/mL (351.67 mM)
SMILES O=C(C=C(C)N1)NC1=S
Molecular Weight (g/mol) 142.18
Formula Weight 142.18
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.