missing translation for 'onlineSavingsMsg'
Learn More

Methylthiomethyl p-tolyl sulfone, 99%, Thermo Scientific™

Product Code. 15484537
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15484537 5 g 5g
15494537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484537 Supplier Thermo Scientific Alfa Aesar Supplier No. H31877.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59662-65-6
Molecular Formula C9H12O2S2
Molecular Weight (g/mol) 216.313
MDL Number MFCD00010750
InChI Key XAARLLNXZJTFPQ-UHFFFAOYSA-N
Synonym methylthiomethyl p-tolyl sulfone, methyl tosylmethyl sulfane, mt-sulfone, methylmercaptomethyl p-tolyl sulfone, 1-methyl-4-methylsulfanylmethylsulfonyl benzene, 1-methyl-4-methylsulfanyl methanesulfonylbenzene, methylthiomethylp-tolylsulfone, acmc-209mf0, methyl thiomethyl p-tolylsulfone, xaarllnxzjtfpq-uhfffaoysa
PubChem CID 548421
IUPAC Name 1-methyl-4-(methylsulfanylmethylsulfonyl)benzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)CSC

Specifications

Melting Point 77°C to 81°C
Quantity 5 g
Formula Weight 216.32
Percent Purity 99%
Chemical Name or Material Methylthiomethyl p-tolyl sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.