missing translation for 'onlineSavingsMsg'
Learn More

(Methylthio)acetic acid, 98%

Product Code. 11440737
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440737 25 g 25g
11420737 1 g 1g
11430737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440737 Supplier Thermo Scientific Alfa Aesar Supplier No. B22531.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2444-37-3 | C3H6O2S | 106.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2444-37-3
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.139
MDL Number MFCD00075444
InChI Key HGTBAIVLETUVCG-UHFFFAOYSA-N
Synonym methylthio acetic acid, 2-methylthioacetic acid, 2-methylthio acetic acid, acetic acid, methylthio, 2-methylsulfanyl acetic acid, methylsulfenylacetic acid, unii-umv7e1ucux, methylmercaptoacetic acid, umv7e1ucux, acetic acid,2-methylthio
PubChem CID 75551
ChEBI CHEBI:47870
IUPAC Name 2-methylsulfanylacetic acid
SMILES CSCC(=O)O

Specifications

Melting Point 13°C to 14°C
Density 1.224
Boiling Point 99°C to 100°C (4 mmHg)
Flash Point >110°C (230°F)
Odor Stench
Refractive Index 1.495
Quantity 25 g
Beilstein 1740201
Formula Weight 106.15
Percent Purity 98%
Chemical Name or Material (Methylthio)acetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.