missing translation for 'onlineSavingsMsg'
Learn More

(Methylthio)acetic acid, +98%, ACROS Organics™

Product Code. 10095453 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10095453 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10095453 Supplier Thermo Scientific Acros Supplier No. 376090250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2444-37-3
CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2190g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C3H6O2S
Linear Formula CH3SCH2CO2H
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
MDL Number MFCD00075444
Quantity 25 mL
Synonym methylthio acetic acid, 2-methylthioacetic acid, 2-methylthio acetic acid, acetic acid, methylthio, 2-methylsulfanyl acetic acid, methylsulfenylacetic acid, unii-umv7e1ucux, methylmercaptoacetic acid, umv7e1ucux, acetic acid,2-methylthio
InChI Key HGTBAIVLETUVCG-UHFFFAOYSA-N
SMILES CSCC(=O)O
IUPAC Name 2-methylsulfanylacetic acid
Molecular Weight (g/mol) 106.139
PubChem CID 75551
ChEBI CHEBI:47870
Formula Weight 106.14
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material (Methylthio)acetic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.